
Glucagon-Like Peptide I Amide Fragment 7-36 human

Synonym(s):
Preproglucagon 78-107 Amide
Empirical Formula (Hill Notation):
C149H226N40O45
CAS Number:
Molecular Weight:
3297.63
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing