
Geniposidic acid

Synonym(s):
(1S,4aS,7aS)-1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)-cyclopenta[c]pyran-4-carboxylic acid
Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing