![Fenbendazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/526/09ce670c-19ab-45ce-83af-19f8e5b408db/640/09ce670c-19ab-45ce-83af-19f8e5b408db.png)
Fenbendazole impurity A
![Fenbendazole impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/526/09ce670c-19ab-45ce-83af-19f8e5b408db/640/09ce670c-19ab-45ce-83af-19f8e5b408db.png)
Synonym(s):
Carbendazim, BCM, Methyl 2-benzimidazolecarbamate, Methyl benzimidazol-2-ylcarbamate
Empirical Formula (Hill Notation):
C9H9N3O2
CAS Number:
Molecular Weight:
191.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing