Skip to Content
Merck
Coumarin 314 Dye content 97 %

Coumarin 314

Coumarin 314 Dye content 97 %
Synonym(s):
2,3,5,6-1H,4H-Tetrahydro-9-carbethoxyquinolizino-[9,9a,1-gh]coumarin, Coumarin 504
Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing