
Asperulosidic acid

Empirical Formula (Hill Notation):
C18H24O12
CAS Number:
Molecular Weight:
432.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing