Skip to Content
Merck
All Photos(1)

Key Documents

90649

Supelco

1H,1H-Heptafluoro-1-butanol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,2,3,3,4,4,4-Heptafluoro-1-butanol, Perfluoropropyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2CH2OH
CAS Number:
Molecular Weight:
200.05
Beilstein:
1761907
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

LC/MS: suitable

refractive index

n20/D 1.3 (lit.)

bp

96-97 °C (lit.)

density

1.6 g/mL at 25 °C (lit.)

application(s)

clinical testing
coatings
environmental
food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4H3F7O/c5-2(6,1-12)3(7,8)4(9,10)11/h12H,1H2

InChI key

WXJFKAZDSQLPBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service