Merck
HomeWestern BlottingImmobilon® PVDF membranes for Western blotting
Immobilon® PVDF membranes for Western blotting
How do Western blot membranes work?
Polyvinylidene fluoride (PVDF) membranes bind biomolecules through hydrophobic interactions. Membrane pores increase the surface binding area while restricting sizes of bound proteins.
We offer four types of Immobilon® PVDF (Polyvinylidene Difluoride) membranes, each optimized for a specific protein blotting application. Blotting sandwiches that feature pre-cut sheets of membrane and blotting filter paper are also available.
  • Immobilon®-E membrane is the only PVDF membrane (0.45 µM) that wets out in aqueous buffers, eliminating the methanol pre-wet step. This membrane is well-suited for most Western blotting applications.
  • Immobilon®-P membrane is optimized for Western blotting and offer better handling and staining than nitrocellulose. All Immobilon®-P PVDF membranes have a pore size of 0.45 μm.
  • Immobilon®-PSQ membrane (0.2 µM) is ideal for protein sequencing and immunoblotting of low molecular weight proteins. It has a higher protein binding capacity and a higher retention than 0.45 µM membranes.
  • Immobilon®-FL membrane (0.45 µM) was developed for fluorescence-based immunodetection. It has low background fluorescence across a wide range of excitation and emission wavelengths.
  • Immobilon® NOW membrane has convenient pre‑cut sheets with the flexibility of rolls in each format: Immobilon®-E, Immobilon®-P, Immobilon®-FL and Immobilon®-PSQ membranes.
PVDF Western blot membrane advantages compared to nitrocellulose
  • Won’t crack or curl, and they can be cut without fracturing.
  • Low background
  • Broad solvent compatibility
  • Superior staining capabilities
  • Can be reprobed multiple times
  • Compatible with a variety of detection chemistries, including radioactive, chromogenic, chemiluminescent, fluorescent, and chemifluorescent techniques
Immobilon® PVDF membranes offer excellent protein binding and high signal.
Immobilon® PVDF membranes have high protein adsorption, so you won’t lose proteins during transfer or reprobing. The open pore structure makes it easy to access bound proteins and remove unbound probes. In addition, Immobilon® PVDF membranes optimized for fluorescent blots dramatically increase signal for high sensitivity in quantitative, multiplexing applications.
PVDF Membranes
Membrane
Name
Immobilon®-E membraneImmobilon®-P membraneImmobilon®-FL membraneImmobilon®-PSQ membrane
Pore Size0.45 µM 0.45 µM0.45 µM 0.2 µM
Detection
methods
Chemiluminescence, ColorimetricChemiluminescence, ColorimetricChemililuminescence, Colorimetric, Fluorescence (including near-infrared)Chemiluminescence, Colorimetric
Binding capacityGoat IgG225-293 µg/cm2294 µg/cm2300 µg/cm2448 µg/cm2
BSAn/a215 µg/cm2205 µg/cm2340 µg/cm2
Insulinn/a160 µg/cm2155 µg/cm2262 µg/cm2
ApplicationsWestern blotsWestern blots, dot blotsWestern blots, dot blotsWestern blots, dot blots
The durability and high protein retention of Immobilon® PVDF membranes make them
the preferred option for sequential detection of multiple targets via antibody stripping
and reprobing.
Selection
Criteria
  • Target protein expressed over a broad dynamic range
  • Wide range of molecular weights
  •  Target protein
    <20 kDa
  • Low-abundance target protein
  • Low-affinity antibody