HPLC Analysis of Mianserin Enantiomers on Astec® CHIROBIOTIC® V
![HPLC Analysis of Mianserin Enantiomers on Astec® CHIROBIOTIC® V application for HPLC](/deepweb/assets/sigmaaldrich/product/images/378/116/0f03cf0a-74e6-4ed0-939f-5aebb3b30be7/800/0f03cf0a-74e6-4ed0-939f-5aebb3b30be7.jpg)
Materials
analytical column
Product No.
Description
Pricing
Application
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate
1 mL/min
column temp.
29 °C
detector
UV, 254 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC