HPLC Analysis of Carboxymethylated Apohemoglobin Tryptic Digest on Discovery® BIO Wide Pore C18
![HPLC Analysis of Carboxymethylated Apohemoglobin Tryptic Digest on Discovery® BIO Wide Pore C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/232/147/c3ea9590-fbf6-4050-b713-20f8c3f1edf4/800/c3ea9590-fbf6-4050-b713-20f8c3f1edf4.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 4.6 mm I.D., 5 μm particles (568222-U)
mobile phase
[A] (0.1% TFA in water):(0.1% TFA in acetonitrile) 95:5; [B] (0.1% TFA in water):(0.1% TFA in acetonitrile):50:50;
gradient
0-100% B in 65 min
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
50 μL
sample
carboxymethylated apohemoglobin tryptic digest in 50 mM ammonium bicarbonate
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany