Merck
GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX B-DA
GC Analysis of Methylphenidate (Ritalin) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-DA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-DA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
Methylphenidate hydrochloride
CONDITIONS
column
Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 μm (72023AST)
oven
170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 35 psi
sample
peaks 1 & 2: methylphenidate enantiomers (N-TFA derivative)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC