GC Analysis of Blood Alcohols on SUPELCOWAX 10
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOWAX 10, 30 m × 0.25 mm I.D., 0.50 μm (24284)
oven
35 °C (1 min), 10 °C/min to 125 °C (1 min)
detector
FID, 200 °C
carrier gas
helium, 1.0 mL/min constant
injection
0.5 μL, 100:1 split
liner
4 mm I.D. split, cup design
sample
blood alcohols, each analyte at 0.08% in water
Description
Legal Information
null