Merck
  • Enhanced glucose cycling and suppressed de novo synthesis of glucose-6-phosphate result in a net unchanged hepatic glucose output in ob/ob mice.
Enhanced glucose cycling and suppressed de novo synthesis of glucose-6-phosphate result in a net unchanged hepatic glucose output in ob/ob mice.
Diabetologia (2004-12-16)
R H J Bandsma, A Grefhorst, T H van Dijk, F H van der Sluijs, A Hammer, D-J Reijngoud, F Kuipers
ABSTRACT
Leptin-deficient ob/ob mice are hyperinsulinaemic and hyperglycaemic; however, the cause of hyperglycaemia remains largely unknown. Glucose metabolism in vivo in 9-h fasted ob/ob mice and lean littermates was studied by infusing [U-(13)C]-glucose, [2-(13)C]-glycerol, [1-(2)H]-galactose and paracetamol for 6 h, applying mass isotopomer distribution analysis on blood glucose and urinary paracetamol-glucuronide. When expressed on the basis of body weight, endogenous glucose production (109+/-23 vs 152+/-27 micromol.kg(-1).min(-1), obese versus lean mice, p<0.01) and de novo synthesis of glucose-6-phosphate (122+/-13 vs 160+/-6 micromol.kg(-1).min(-1), obese versus lean mice, p<0.001) were lower in ob/ob mice than in lean littermates. In contrast, glucose cycling was greatly increased in obese mice (56+/-13 vs 26+/-4 micromol.kg(-1).min(-1), obese versus lean mice, p<0.001). As a result, total hepatic glucose output remained unaffected (165+/-31 vs 178+/-28 micromol.kg(-1).min(-1), obese vs lean mice, NS). The metabolic clearance rate of glucose was significantly lower in obese mice (8+/-2 vs 18+/-2 ml.kg(-1).min(-1), obese versus lean mice, p<0.001). Hepatic mRNA levels of genes encoding for glucokinase and pyruvate kinase were markedly increased in ob/ob mice. Unaffected total hepatic glucose output in the presence of hyperinsulinaemia reflects hepatic insulin resistance in ob/ob mice, which is associated with markedly increased rates of glucose cycling. Hyperglycaemia in ob/ob mice primarily results from a decreased metabolic clearance rate of glucose.