Merck
  • Rhodium-catalyzed direct C-H addition of 3,4-dihydroquinazolines to alkenes and their use in the total synthesis of vasicoline.
Rhodium-catalyzed direct C-H addition of 3,4-dihydroquinazolines to alkenes and their use in the total synthesis of vasicoline.
The Journal of organic chemistry (2006-02-25)
Sean H Wiedemann, Jonathan A Ellman, Robert G Bergman
ABSTRACT
The inter- and intramolecular couplings of unactivated alkenes to 3,4-dihydroquinazolines with a Rh(I) catalyst are reported. Coupling between olefins and NH-3,4-dihydroquinazoline was found to occur consecutively with heterocycle dehydrogenation in the presence of a Rh(I)/PCy3/HCl catalyst. The reaction was used to develop an effective method for the synthesis of 2-substituted quinazolines through an oxidative workup step. The regiocontrolled synthesis and Rh-catalyzed cyclization of alkene-tethered 3,4-dihydroquinazolines are also described. Applying this method, the second total synthesis of vasicoline was achieved. The key Rh-catalyzed cyclization step was made possible by the use of a rigid bicyclic phosphine ligand. The synthesis further demonstrates a challenging Cu-catalyzed amidation of an ortho-substituted aryl chloride.
Sigma-Aldrich
DTBPF
Pricing and availability is not currently available.