- Comparative Metagenomics of Cellulose- and Poplar Hydrolysate-Degrading Microcosms from Gut Microflora of the Canadian Beaver (
Comparative Metagenomics of Cellulose- and Poplar Hydrolysate-Degrading Microcosms from Gut Microflora of the Canadian Beaver (
Frontiers in microbiology (2018-01-13)
Mabel T Wong, Weijun Wang, Marie Couturier, Fakhria M Razeq, Vincent Lombard, Pascal Lapebie, Elizabeth A Edwards, Nicolas Terrapon, Bernard Henrissat, Emma R Master
PMID29326667
ABSTRACT
To identify carbohydrate-active enzymes (CAZymes) that might be particularly relevant for wood fiber processing, we performed a comparative metagenomic analysis of digestive systems from Canadian beaver (