Merck
The Intermetalloid Cluster Cation (CuBi
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-10-05)
Maximilian Knies, Martin Kaiser, Anna Isaeva, Ulrike Müller, Thomas Doert, Michael Ruck
ABSTRACT
The reaction of Bi, BiCl
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%