Merck
  • FilaminA and Formin2 regulate skeletal, muscular, and intestinal formation through mesenchymal progenitor proliferation.
FilaminA and Formin2 regulate skeletal, muscular, and intestinal formation through mesenchymal progenitor proliferation.
PloS one (2017-12-15)
Gewei Lian, Sneha Kanaujia, Timothy Wong, Volney Sheen
ABSTRACT
The effects of actin dependent molecular mechanisms in coordinating cellular proliferation, migration and differentiation during embryogenesis are not well-understood. We have previously shown that actin-binding Filamin A (FlnA) and actin-nucleating Formin 2 (Fmn2) influence the development of the brain causing microcephaly in mice. In this study, we broaden this phenotype to explore the effects of these two proteins in the development of extra-CNS organ systems, including the gut, muscle, and skeleton. We observed defects in rib and sternum midline closure leading to thoracoabdominal schisis in FlnA+Fmn2 knockout mice, reminiscent of the pentalogy of Cantrell syndrome. These mice exhibit shortened guts, as well as thinned thoracic muscle mass. Immunostaining showed these changes are partially caused by a decrease in the number of presumptive mesenchymal proliferating cells with loss of either FlnA or FlnA+Fmn2. This proliferation defect appears to be in part due to delayed differentiation in these regions. While both FlnA and FlnA+Fmn2 mice show reduced cell death relative to WT control, increased caspase staining was seen in the double null relative to FlnA null suggesting that this could also contribute to the FlnA+Fmn2 phenotype. Therefore FlnA and Fmn2 are likely essential to cell proliferation, differentiation and cell death in a variety of tissues and organs, further reiterating the importance of vesicle trafficking in regulation of development.