Merck
  • A positive circuit of VEGF increases Glut-1 expression by increasing HIF-1α gene expression in human retinal endothelial cells.
A positive circuit of VEGF increases Glut-1 expression by increasing HIF-1α gene expression in human retinal endothelial cells.
Archives of pharmacal research (2017-10-13)
Yoon Kyung Choi
ABSTRACT
Treatment of human retinal microvascular endothelial cells (HRMECs) with vascular endothelial growth factor 165 (VEGF
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKCG
Sigma-Aldrich
2-Phenylindole, technical grade, 95%