Merck
  • Nuclear IGF-1R predicts chemotherapy and targeted therapy resistance in metastatic colorectal cancer.
Nuclear IGF-1R predicts chemotherapy and targeted therapy resistance in metastatic colorectal cancer.
British journal of cancer (2017-11-11)
Jordi Codony-Servat, Miriam Cuatrecasas, Elena Asensio, Carla Montironi, Anna Martínez-Cardús, Mercedes Marín-Aguilera, Carlos Horndler, Eva Martínez-Balibrea, Michele Rubini, Pedro Jares, Oscar Reig, Iván Victoria, Lydia Gaba, Marta Martín-Richard, Vicente Alonso, Pilar Escudero, Carlos Fernández-Martos, Jaime Feliu, Jose Carlos Méndez, Miguel Méndez, Javier Gallego, Antonieta Salud, Federico Rojo, Antoni Castells, Aleix Prat, Rafael Rosell, Xabier García-Albéniz, Jordi Camps, Joan Maurel
ABSTRACT
Although chemotherapy is the cornerstone treatment for patients with metastatic colorectal cancer (mCRC), acquired chemoresistance is common and constitutes the main reason for treatment failure. Monoclonal antibodies against insulin-like growth factor-1 receptor (IGF-1R) have been tested in pre-treated mCRC patients, but results have been largely deceiving. We analysed time to progression, overall survival, and the mutational status of RAS, BRAF and nuclear p-IGF-1R expression by immunohistochemistry, in 470 metastatic CRC patients. The effect of IGF-1R activation and distribution was also assessed using cellular models of CRC and RNAi for functional validation. Nuclear IGF-1R increased in metastatic tumours compared to paired untreated primary tumours, and significantly correlated with poor overall survival in mCRC patients. In vitro, chemo-resistant cell lines presented significantly higher levels of IGF-1R expression within the nuclear compartment, and PIAS3, a protein implicated also in the sumoylation process of intranuclear proteins, contributed to IGF-1R nuclear sequestration, highlighting the essential role of PIAS3 in this process. Intriguingly, we observed that ganitumab, an IGF-1R blocking-antibody used in several clinical trials, and dasatinib, an SRC inhibitor, increased the nuclear localisation of IGF-1R. Our study demonstrates that IGF-1R nuclear location might lead to chemotherapy and targeted agent resistance.
Sigma-Aldrich
Dasatinib
Sigma-Aldrich
MISSION® esiRNA, targeting human PIAS3
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)