Merck
Pressure dependence of side chain
Journal of biomolecular NMR (2017-09-16)
Markus Beck Erlach, Joerg Koehler, Edson Crusca, Claudia E Munte, Masatsune Kainosho, Werner Kremer, Hans Robert Kalbitzer
ABSTRACT
For evaluating the pressure responses of folded as well as intrinsically unfolded proteins detectable by NMR spectroscopy the availability of data from well-defined model systems is indispensable. In this work we report the pressure dependence of
Sigma-Aldrich
Fmoc-Leu-OH, ≥97.0%