Merck
  • Antitumor Activity of the Chinese Medicine JC-001 Is Mediated by Immunomodulation in a Murine Model of Hepatocellular Carcinoma.
Antitumor Activity of the Chinese Medicine JC-001 Is Mediated by Immunomodulation in a Murine Model of Hepatocellular Carcinoma.
Integrative cancer therapies (2016-10-05)
Meng-Hsien Chuang, Jinghua Tsai Chang, Li-Jin Hsu, Ming-Shiou Jan, Fung-Jou Lu
ABSTRACT
JC-001 is a Chinese medicine that has been used to treat liver disease; however, its significance in cancer treatment has not been characterized. In this study, we used an immunocompetent tumor model to characterize the antitumor activity of JC-001. A total of 48 Hepa 1-6 tumor-bearing C57BL/6 mice were randomly grouped into 4 groups and treated with H
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)