Merck
Type 1 diabetes induction in humanized mice.
Proceedings of the National Academy of Sciences of the United States of America (2017-09-07)
Shulian Tan, Yang Li, Jinxing Xia, Chun-Hui Jin, Zheng Hu, Gaby Duinkerken, Yuying Li, Mohsen Khosravi Maharlooei, Estefania Chavez, Grace Nauman, Nichole Danzl, Maki Nakayama, Bart O Roep, Megan Sykes, Yong-Guang Yang
ABSTRACT
There is an urgent and unmet need for humanized in vivo models of type 1 diabetes to study immunopathogenesis and immunotherapy, and in particular antigen-specific therapy. Transfer of patient blood lymphocytes to immunodeficient mice is associated with xenogeneic graft-versus-host reactivity that complicates assessment of autoimmunity. Improved models could identify which human T cells initiate and participate in beta-cell destruction and help define critical target islet autoantigens. We used humanized mice (hu-mice) containing robust human immune repertoires lacking xenogeneic graft-versus-host reactivity to address this question. Hu-mice constructed by transplantation of HLA-DQ8
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%