Merck
Gas-Phase Reaction of Methyl n-Propyl Ether with OH, NO
The journal of physical chemistry. A (2017-08-25)
Jianqiang Zhu, Shuyan Wang, Narcisse T Tsona, Xiaotong Jiang, Yifeng Wang, Maofa Ge, Lin Du
ABSTRACT
Rate constants at room temperature (293 ± 2 K) and atmospheric pressure for the reaction of methyl n-propyl ether (MnPE), CH
Sigma-Aldrich
Trichloroacetyl chloride, 99%