Merck
  • Functional modification of breathable polyacrylonitrile/polyurethane/TiO
Functional modification of breathable polyacrylonitrile/polyurethane/TiO
Journal of colloid and interface science (2017-09-04)
Yue Xu, Junlu Sheng, Xia Yin, Jianyong Yu, Bin Ding
ABSTRACT
Exploiting high-added-value textiles equipped with multiple functionalities like ultraviolet (UV) resistance, waterproofness, and thermal-moisture comfort is facing tremendous demand by a more discerning consumer market. However, the major challenge is to realize the equilibrium among the multifunction. Herein, a new attempt of fabricating superhydrophobic electrospun polyacrylonitrile (PAN)/polyurethane (PU)/titanium dioxide (TiO
Sigma-Aldrich
2-Hydroxy-4-(octyloxy)benzophenone, 98%