MilliporeSigma
  • Living with a leaky skin: upregulation of ion transport proteins during sloughing.
Living with a leaky skin: upregulation of ion transport proteins during sloughing.
The Journal of experimental biology (2017-06-02)
Nicholas C Wu, Rebecca L Cramp, Craig E Franklin
ABSTRACT
Amphibian skin is a multifunctional organ providing protection from the external environment and facilitating the physiological exchange of gases, water and salts with the environment. In order to maintain these functions, the outer layer of skin is regularly replaced in a process called sloughing. During sloughing, the outermost layer of the skin is removed in its entirety, which has the potential to interfere with skin permeability and ion transport, disrupting homeostasis. In this study, we measured, in vivo, the effects of sloughing on the cutaneous efflux of ions in toads Rhinella marina kept in freshwater conditions. We also measured transepithelial potential, cutaneous resistance, active ion transport and the distribution, abundance and gene expression of the key ion transport proteins sodium-potassium ATPase (NKA) and epithelial sodium channel (ENaC) during sloughing. We hypothesised that the increase in transepithelial efflux of ions during sloughing is a consequence of increased permeability and/or a reduction in the abundance or expression of cutaneous ion transport proteins, resulting in disruption of internal ion homeostasis. There was a significant increase in sodium and chloride efflux during sloughing in R. marina However, although in vitro skin resistance decreased after sloughing, active sodium transport increased commensurate with an increase in NKA and ENaC protein abundance in the skin. These changes in skin function associated with sloughing did not affect the maintenance of internal electrolyte homeostasis. These results suggest that during sloughing, amphibians actively maintain internal homeostasis by increasing cutaneous rates of ion uptake.