Merck
Identity and function of a cardiac mitochondrial small conductance Ca
Biochimica et biophysica acta (2017-03-28)
MeiYing Yang, Amadou K S Camara, Mohammed Aldakkak, Wai-Meng Kwok, David F Stowe
ABSTRACT
We provide evidence for location and function of a small conductance, Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human KCNN3