Merck
Catalytic topological insulator Bi
Nanomedicine : nanotechnology, biology, and medicine (2017-03-13)
Xiao-Dong Zhang, Yaqi Jing, Shasha Song, Jiang Yang, Jun-Ying Wang, Xuhui Xue, Yuho Min, Gyeongbae Park, Xiu Shen, Yuan-Ming Sun, Unyong Jeong
ABSTRACT
Bi
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, ≥99.99% trace metals basis