Merck
  • Timing and duration of nursing from birth affect neonatal porcine uterine matrix metalloproteinase 9 and tissue inhibitor of metalloproteinase 1.
Timing and duration of nursing from birth affect neonatal porcine uterine matrix metalloproteinase 9 and tissue inhibitor of metalloproteinase 1.
Domestic animal endocrinology (2016-11-21)
T Y Ho, K M Rahman, M E Camp, A A Wiley, F F Bartol, C A Bagnell
ABSTRACT
Nursing for 2 d from birth supports neonatal porcine uterine and cervical development. However, it is not clear how timing or duration of lactocrine signaling from birth (postnatal day = PND 0) affects development of neonatal female reproductive tract tissues. Therefore, studies were conducted to determine effects of age at first nursing and duration of nursing from birth on specific elements of the matrix metalloproteinase (MMP)/tissue inhibitor of metalloproteinase (TIMP) system in uterine and cervical tissues at PND 2. When nursing was initiated at 0 h or 30 min of age, targeted proteins, including proMMP9 and MMP9, were detected in uterine and cervical tissues on PND 2, as was uterine TIMP1. However, these proteins were undetectable when nursing was delayed for 12 h and when gilts were fed milk replacer for 48 h from birth. Increasing the duration of nursing from 30 min to 12 h from birth increased uterine (P < 0.05) and cervical (P < 0.001) MMP9 levels to those observed in gilts nursed for 48 h. Similarly, uterine TIMP1 levels increased with duration of nursing. Uterine MMP2 levels were detectable but unaffected by age at first nursing or duration of nursing from birth. Uterine MMP2 and MMP9 activities, monitored by zymography, reflected immunoblotting data. Results provide evidence for the utility of MMP9 and TIMP1 as markers of age- and lactocrine-sensitive porcine female reproductive tract development.
Sigma-Aldrich
Anti-MMP-2 (Ab-3) Mouse mAb (42-5D11), liquid, clone 42-5D11, Calbiochem®
Sigma-Aldrich
TIMP1 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)