Merck
Novel ion exchange chromatography method for nca arsenic separation.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2017-01-29)
Z Oláh, T Kremmer, A T Vogg, Z Varga, Z Szűcs, B Neumaier, R Dóczi
ABSTRACT
A high-performance liquid chromatography (HPLC) device equipped with an anion exchange column was used to isolate nca
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis