Merck
  • Increased Dynamics of Tricarboxylic Acid Cycle and Glutamate Synthesis in Obese Adipose Tissue: IN VIVO METABOLIC TURNOVER ANALYSIS.
Increased Dynamics of Tricarboxylic Acid Cycle and Glutamate Synthesis in Obese Adipose Tissue: IN VIVO METABOLIC TURNOVER ANALYSIS.
The Journal of biological chemistry (2017-01-26)
Hirofumi Nagao, Hitoshi Nishizawa, Takeshi Bamba, Yasumune Nakayama, Noriyoshi Isozumi, Shushi Nagamori, Yoshikatsu Kanai, Yoshimitsu Tanaka, Shunbun Kita, Shiro Fukuda, Tohru Funahashi, Norikazu Maeda, Eiichiro Fukusaki, Iichiro Shimomura
ABSTRACT
Obesity is closely associated with various metabolic disorders. However, little is known about abnormalities in the metabolic change of obese adipose tissue. Here we use static metabolic analysis and
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC1A3