- Droplet Assisted Inlet Ionization for Online Analysis of Airborne Nanoparticles.
Droplet Assisted Inlet Ionization for Online Analysis of Airborne Nanoparticles.
Airborne nanoparticles play a key role in climate effects as well as impacting human health. Their small mass and complex chemical composition represent significant challenges for analysis. This work introduces a new ionization method, droplet assisted inlet ionization (DAII), where aqueous droplets are produced from airborne nanoparticles. When these droplets enter the mass spectrometer through a heated inlet, rapid vaporization leads to the formation of molecular ions. The method is demonstrated with test aerosols consisting of polypropylene glycol (PPG), angiotensin II, bovine serum albumin, and the "thermometer" compound p-methoxybenzylpyridinium chloride. High-quality spectra were obtained from PPG particles down to 13 nm in diameter and sampled masses in the low pictogram range. These correspond to aerosol number and mass concentrations smaller than 1000 particles/cm