Merck
  • Naked eye and spectrophotometric detection of chromogenic insecticide in aquaculture using amine functionalized gold nanoparticles in the presence of major interferents.
Naked eye and spectrophotometric detection of chromogenic insecticide in aquaculture using amine functionalized gold nanoparticles in the presence of major interferents.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-11-07)
C Loganathan, S Abraham John
ABSTRACT
Detection of a chromogenic insecticide, malachite green (MG) using 3,5-diamino-1,2,4-triazole capped gold nanoparticles (DAT-AuNPs) by both naked eye and spectrophotometry was described in this paper. The DAT-AuNPs were prepared by wet chemical method and show absorption maximum at 518nm. The zeta potential of DAT-AuNPs was found to be -39.9mV, suggesting that one of the amine groups of DAT adsorbed on the surface of AuNPs and the other amine group stabilizes the AuNPs from aggregation. The wine red color DAT-AuNPs changes to violet while adding 25μM MG whereas the absorption band at 518nm was increased and shifted towards longer wavelength. However, addition of 70μM MG leads to the aggregation of DAT-AuNPs. This is due to strong electrostatic interaction between ammonium ion of MG and the free amine group of DAT. Based on the color change and shift in SPR band, 25 and 5μM MG can be easily detected by naked eye and spectrophotometry. The DAT-AuNPs show high selectivity towards MG even in the presence of 5000-fold higher concentrations of common interferents. The practical application was successfully demonstrated by determining MG in fish farm water.
Sigma-Aldrich
3,5-Diamino-1,2,4-triazole, 98%