Merck
  • Revisiting catechol derivatives as robust chromogenic hydrogen donors working in alkaline media for peroxidase mimetics.
Revisiting catechol derivatives as robust chromogenic hydrogen donors working in alkaline media for peroxidase mimetics.
Analytica chimica acta (2016-11-23)
Marcin Drozd, Mariusz Pietrzak, Jakub Pytlos, Elżbieta Malinowska
ABSTRACT
Colloidal noble metal-based nanoparticles are able to catalyze oxidation of chromogenic substrates by H