Merck
  • Efficient Differentiation of Human Induced Pluripotent Stem Cell (hiPSC) Derived Hepatocyte-Like Cells on hMSCs Feeder.
Efficient Differentiation of Human Induced Pluripotent Stem Cell (hiPSC) Derived Hepatocyte-Like Cells on hMSCs Feeder.
International journal of hematology-oncology and stem cell research (2015-03-17)
Naser Mobarra, Masoud Soleimani, Fatemeh Kouhkan, Zahra Hesari, Reyhaneh Lahmy, Majid Mossahebi-Mohammadi, Ehsan Arefian, Zahra Jaafarpour, Hajar Nasiri, Reza Pakzad, Rezvan Tavakoli, Parvin Pasalar
ABSTRACT
The use of stem cells is considered as an appropriate source in cell therapy and tissue engineering. Differentiation of human induced Pluripotent Stem Cells (hiPSCs) to Hepatocyte-like Cells (HLCs) on mouse embryonic fibroblasts (MEFs) feeders is confronted with several problems that hinder the clinical applications of these differentiated cells for the treatment of liver injuries. Safe appropriate cells for stem cell-based therapies could create new hopes for liver diseases. This work focused on the determination of a capacity/efficiency for the differentiation of the hiPSCs into Hepatocyte-like Cells on a novel human adult bone marrow mesenchymal stem cells (hMSCs) feeder. Undifferentiated human iPSCs were cultured on mitotically inactivated human adult bone marrow mesenchymal stem cells. A three-step differentiation process has been performed in presence of activin A which added for 3 days to induce a definitive endoderm formation. In the second step, medium was exchanged for six days. Subsequently, cells were treated with oncostatin M plus dexamethasone for 9 days to generate hepatic cells. Endodermic and liver-specific genes were assessed via quantitative reverse transcription-polymerase chain reaction and RT-PCR, moreover, immunocytochemical staining for liver proteins including albumin and alpha-fetoprotein. In addition, functional tests for glycogen storage, oil red examination, urea production and alpha-fetoprotein synthesis, as well as, cells differentiated with a hepatocyte-like morphology was also performed. Our results show that inactivated human adult bone marrow mesenchymal stem cell feeders could support the efficient differentiation of hiPSCs into HLCs. This process induced differentiation of iPSCs into definitive endocrine cells that expressed sox17, foxa2 and expression of the specific genes profiles in hepatic-like cells. In addition, immunocytochemical analysis confirmed albumin and alpha-fetoprotein protein expression, as well as, the hiPSCs-derived Hepatocyte-like Cells on human feeder exhibited a typical morphology. we suggested a successful and efficient culture for differentiation and maturation of hepatocytes on an alternative human feeders; this is an important step to generate safe and functional hepatocytes that is vital for regenerative medicine and transplantation on the cell-based therapies.