Merck
  • Chromosomal assignment of a human apoptosis-associated tyrosine kinase gene on chromosome 17q25.3 by somatic hybrid analysis and fluorescence in situ hybridization.
Chromosomal assignment of a human apoptosis-associated tyrosine kinase gene on chromosome 17q25.3 by somatic hybrid analysis and fluorescence in situ hybridization.
Journal of human genetics (1999-03-20)
N Seki, A Hayashi, A Hattori, S Kozuma, M Ohira, T Hori, T Saito
ABSTRACT
Here, we report on the chromosomal location of the human apoptosis-associated tyrosine kinase gene. Based on polymerase chain reaction analysis with a human/rodent monochromosomal hybrid cell panel and fluorescence in situ hybridization, the gene was mapped on 25.3 region of chromosome 17.