Merck
  • A novel chromosome 19p13.12 deletion in a child with multiple congenital anomalies.
A novel chromosome 19p13.12 deletion in a child with multiple congenital anomalies.
American journal of medical genetics. Part A (2009-02-14)
Daniel R Jensen, Donna M Martin, Stephen Gebarski, Trilochan Sahoo, Ellen K Brundage, A Craig Chinault, Edgar A Otto, Moumita Chaki, Friedhelm Hildebrandt, Sau Wai Cheung, Marci M Lesperance
ABSTRACT
We describe a patient with multiple congenital anomalies including deafness, lacrimal duct stenosis, strabismus, bilateral cervical sinuses, congenital cardiac defects, hypoplasia of the corpus callosum, and hypoplasia of the cerebellar vermis. Mutation analysis of EYA1, SIX1, and SIX5, genes that underlie otofaciocervical and/or branchio-oto-renal syndrome, was negative. Pathologic diagnosis of the excised cervical sinus tracts was revised on re-examination to heterotopic salivary gland tissue. Using high resolution chromosomal microarray analysis, we identified a novel 2.52 Mb deletion at 19p13.12, which was confirmed by fluorescent in situ hybridization and demonstrated to be a de novo mutation by testing of the parents. Overall, deletions of chromosome 19p13 are rare.