Merck
  • beta-Phenylethylamine regulation of dopaminergic nigrostriatal cell activity.
beta-Phenylethylamine regulation of dopaminergic nigrostriatal cell activity.
Brain research (1995-12-12)
M Rodriguez, N Barroso
ABSTRACT
In the present paper, the action of beta-phenylethylamine on electrophysiological activity of dopaminergic nigrostriatal neurons is described. 10 s after its i.v. injection and during 2-4 min, beta-phenylethylamine decreased the firing rate, the number of spikes within and out of burst and the number of bursts per second of these neurons. This was a dose-related action with statistical differences starting from 1.4 mg/kg for total and out of burst firing rate and from 2.4 mg/kg for within burst firing rate and for the number of bursts per second. The standard deviation and the variation coefficient of inter-spike intervals increased in a dose-related way. The marked effect found after low-dose administration suggests that under physiological conditions endogenous beta-phenylethylamine levels regulate the nigrostriatal dopaminergic cell activity. After peripheral low dose administration, beta-phenylethylamine behaves as a dopaminergic agonist with a very fast and brief action.