Merck
  • Plasma antioxidants are similarly depleted in mild cognitive impairment and in Alzheimer's disease.
Plasma antioxidants are similarly depleted in mild cognitive impairment and in Alzheimer's disease.
Neurobiology of aging (2003-08-21)
P Rinaldi, M C Polidori, A Metastasio, E Mariani, P Mattioli, A Cherubini, M Catani, R Cecchetti, U Senin, P Mecocci
ABSTRACT
In order to assess peripheral levels and activities of a broad spectrum of non-enzymatic and enzymatic antioxidants in elderly subjects with mild cognitive impairment (MCI) and Alzheimer's disease (AD), plasma levels of water-soluble (Vitamin C and uric acid) and of lipophilic (Vitamin A, Vitamin E and carotenoids including lutein, zeaxanthin, beta-cryptoxanthin, lycopene, alpha- and beta-carotene) antioxidant micronutrients as well as activities of plasma and red blood cell (RBC) superoxide dismutase (SOD) and of plasma glutathione peroxidase (GPx) were measured in 25 patients with MCI, 63 AD patients and 53 controls. Peripheral levels and activities of antioxidants were similarly lower in MCI and AD patients as compared to controls. As MCI may represent a prodromal stage of AD, and oxidative damage appears to occur as one of the earliest pathophysiological events in AD, an increased intake of antioxidants in patients with MCI could be helpful in lowering the risk of conversion to dementia.
Sigma-Aldrich
(±)-α-Carotene, ≥95.0% (HPLC)