Merck
Kipukasins, nucleoside derivatives from Aspergillus versicolor.
Journal of natural products (2007-07-05)
Ping Jiao, Sanjay V Mudur, James B Gloer, Donald T Wicklow
ABSTRACT
Seven new aroyl uridine derivatives (kipukasins A-G; 1-7) were isolated from solid-substrate fermentation cultures of two different Hawaiian isolates of Aspergillus versicolor. The structures of compounds 1-7 were determined by analysis of NMR and MS data. The nucleoside portion of lead compound 1 was assigned as uracil-1-beta-D-ribofuranoside by spectral comparison with an authentic standard. The bioactivity of the original A. versicolor extracts was accounted for mainly by the presence of the known metabolite sterigmatocystin, but kipukasins A and B showed modest activity in assays against Gram-positive bacteria.
Sigma-Aldrich
Kipukasin A, ≥95% (LC/MS-UV)
Sigma-Aldrich
Kipukasin D, ≥90% (LC/MS-UV)