Merck
  • Toxic effects of mildly elevated homocysteine concentrations in neuronal-like cells.
Toxic effects of mildly elevated homocysteine concentrations in neuronal-like cells.
Neurochemical research (2014-05-29)
M Currò, A Gugliandolo, C Gangemi, R Risitano, R Ientile, D Caccamo
ABSTRACT
Epidemiological and experimental evidence indicated that hyperhomocysteinemia is associated with neurodegeneration. However, homocysteine neurotoxic effects have been so far investigated mostly by employing homocysteine concentrations (≥100 µM) much higher than homocysteine mean plasma levels (20 µM) observed in patients with neurodegenerative disorders. While evaluating the effects of a prolonged exposure to ~20 µM homocysteine in neuronal-like differentiated SH-SY5Y cells, we observed a 35% loss of cell viability and a four-fold increase in reactive oxygen species levels in cells incubated with homocysteine for five days compared with controls. Moreover, homocysteine increased by 30% and around two-fold, respectively, the Comet-positive cell number and DNA damage indexes (tail length, T-DNA, olive tail moment) compared with controls. Cell response to homocysteine-induced DNA damage involved the up-regulation of Bax and, at a greater extent, Bcl-2, but not caspase-3, in association with a p53-independent increase of p21 levels; concomitantly, also p16 levels were increased. When looking at time-dependent changes in cyclin expression, we found that a significant up-regulation of cyclins D1, A1, E1, but not B1, concomitant with p21 down-regulation, occurred in cells incubated with homocysteine for three days. However, in line with the observed increase of p21 and p16 levels, a five days incubation with homocysteine induced cyclin down-regulation accompanied by a strong reduction of phosphorylated pRB amounts. These results suggest that, when prolonged, the exposure of neuronal-like cells to mildly elevated homocysteine concentrations triggers oxidative and genotoxic stress involving an early induction of cyclins, that is late repressed by G1-S check-point regulators.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
5-Methylthiazole, 97%