Merck
  • Magnolol reduces UVB-induced photodamage by regulating matrix metalloproteinase activity.
Magnolol reduces UVB-induced photodamage by regulating matrix metalloproteinase activity.
Environmental toxicology and pharmacology (2015-01-07)
A-Rang Im, Jae Hyoung Song, Mi Young Lee, Sungwook Chae
ABSTRACT
In this study, we evaluated the anti-photoaging activity of magnolol in UV-irradiated hairless mice, and hypothesized that magnolol would prevent photoaging in these animals. The inhibitory effect of magnolol on wrinkle formation was determined by analyzing the skin replica, histologically examining the epidermal thickness, and identifying damage to the collagen fibers. The protective effects of magnolol on UVB-induced skin photoaging were examined by determining the level of MMPs and mitogen-activated protein kinases (MAPKs). Exposure to UVB radiation significantly increased skin thickness and wrinkle grade, but magnolol treatment significantly reduced the average length and depth of wrinkles, and this was correlated with the inhibition of collagen fiber loss. The magnolol-treated group had remarkably decreased activity levels of MMP-1, -9, and -13 compared to the corresponding levels in the vehicle-treated UVB-irradiated group. These results indicate that magnolol prevents skin photoaging in UVB-irradiated hairless mice.
Sigma-Aldrich
Magnolol, ≥90% (HPLC), from plant
Magnolol, European Pharmacopoeia (EP) Reference Standard