Merck
  • c-Src and Pyk2 protein tyrosine kinases play protective roles in early HIV-1 infection of CD4+ T-cell lines.
c-Src and Pyk2 protein tyrosine kinases play protective roles in early HIV-1 infection of CD4+ T-cell lines.
Journal of acquired immune deficiency syndromes (1999) (2014-01-15)
Stephen D S McCarthy, Daniel Jung, Darinka Sakac, Donald R Branch
ABSTRACT
During early HIV-1 infection of CD4 T-lymphocytes, many host protein tyrosine kinases become activated within minutes, including phosphoprotein pp60 (c-Src) and the focal adhesion kinase family member, proline-rich tyrosine kinase 2 (Pyk2). Whether their activation facilitates or impedes infection remains to be determined. c-Src kinase inhibitors (SU6656, PP1, and PP2), adenovectors [wild-type and dominant-negative (DN) c-src] or siRNA (targeting c-src or pyk2) were used to inhibit, compete with or knockdown c-Src in Jurkat C, Jurkat E6-1, Hut 78 or Kit225 T-cell lines. Cells were then infected with HIV-1 luciferase reporter virus expressing VSV-G or HXB2(X4) envelope, and luciferase activity was measured after 2 days. Reverse transcriptase activity and viral cDNA were measured 1 hour after infection, whereas integrated virus was measured 12 hours after infection. Pretreating Jurkat T-cells with SU6656 led to increased VSV-G luciferase activity. In the adenovector experiments, T-cells overexpressing dominant-negative c-Src, but not wild-type c-Src, showed increased luciferase activity after VSV-G infection. siRNA knockdown of c-Src or Pyk2, followed by HXB2 infection in Jurkat T-cells, lead to increased reverse transcriptase activity, viral cDNA, integrated virus, and increased luciferase activity. Pyk2 is known to interact with c-Src. Thus, Pyk2 activation that coincides with increased c-Src activity during HIV-1 infection could be responsible for c-Src activation. Reduced c-Src activation increases HIV-1 reverse transcription, integration, and/or transcription, suggesting the high c-Src activity seen early in HIV-1 infection may be a cellular response to slow or prevent early infection in CD4 T-cells.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%