Merck
  • Phospholipase C epsilon 1 (PLCE1 rs2274223A>G, rs3765524C>T and rs7922612C>T) polymorphisms and esophageal cancer risk in the Kashmir Valley.
Phospholipase C epsilon 1 (PLCE1 rs2274223A>G, rs3765524C>T and rs7922612C>T) polymorphisms and esophageal cancer risk in the Kashmir Valley.
Asian Pacific journal of cancer prevention : APJCP (2014-06-18)
Manzoor Ahmad Malik, Meenakshi Umar, Usha Gupta, Showkat Ali Zargar, Balraj Mittal
ABSTRACT
Phospholipase C epsilon 1 (PLCE1) encodes a member of the phospholipase family of proteins that play crucial roles in carcinogenesis and progression of several cancers including esophageal cancer (EC). In two large scale genome-wide association studies (GWAS) single nucleotide polymorphisms (SNP, rs2274223A>G, rs3765524C>T) in PLCE1 were identified as novel susceptibility loci of esophageal cancer (EC) in China. The aim of the present study was to investigate this finding in Kashmir Valley, a high risk area. We determined genotypes of three potentially functional SNPs (rs2274223A>G, rs3765524C>T and rs7922612C>T) of PLCE1 in 135 EC patients, and 195 age and gender matched controls in Kashmiri valley by PCR RFLP method. Risk for developing EC was estimated by binary logistic regression using SPSS. The selected PLCE1 polymorphisms did not show independent association with EC. However, the G2274223T3765524T7922612 haplotype was significantly associated with increased risk of EC (OR=2.92; 95% CI=1.30-6.54; p=0.009). Smoking and salted tea proved to be independent risk factors for EC. Genetic variations in PLCE1 modulate risk of EC in the high risk Kashmiri population.