Merck
  • Increase in plasma homocysteine associated with parallel increases in plasma S-adenosylhomocysteine and lymphocyte DNA hypomethylation.
Increase in plasma homocysteine associated with parallel increases in plasma S-adenosylhomocysteine and lymphocyte DNA hypomethylation.
The Journal of biological chemistry (2000-07-08)
P Yi, S Melnyk, M Pogribna, I P Pogribny, R J Hine, S J James
ABSTRACT
S-Adenosylmethionine and S-adenosylhomocysteine (SAH), as the substrate and product of essential cellular methyltransferase reactions, are important metabolic indicators of cellular methylation status. Chronic elevation of SAH, secondary to the homocysteine-mediated reversal of the SAH hydrolase reaction, reduces methylation of DNA, RNA, proteins, and phospholipids. High affinity binding of SAH to the active site of cellular methyltransferases results in product inhibition of the enzyme. Using a sensitive new high pressure liquid chromatography method with coulometric electrochemical detection, plasma SAH levels in healthy young women were found to increase linearly with mild elevation in homocysteine levels (r = 0.73; p < 0.001); however, S-adenosylmethionine levels were not affected. Plasma SAH levels were positively correlated with intracellular lymphocyte SAH levels (r = 0.81; p < 0.001) and also with lymphocyte DNA hypomethylation (r = 0.74, p < 0.001). These results suggest that chronic elevation in plasma homocysteine levels, such as those associated with nutritional deficiencies or genetic polymorphisms in the folate pathway, may have an indirect and negative effect on cellular methylation reactions through a concomitant increase in intracellular SAH levels.
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)