Merck
  • An unprecedented octanuclear copper core with C(3i) symmetry and a paramagnetic ground state.
An unprecedented octanuclear copper core with C(3i) symmetry and a paramagnetic ground state.
Chemical communications (Cambridge, England) (2014-01-25)
Dmytro S Nesterov, Julia Jezierska, Oksana V Nesterova, Armando J L Pombeiro, Andrew Ozarowski
ABSTRACT
A complex of copper, possessing an octanuclear structure of high symmetry, has been prepared by a spontaneous self-assembly reaction of Cu(BF4)2 with N-tert-butyldiethanolamine and reveals a paramagnetic ground state.
Sigma-Aldrich
N-tert-Butyldiethanolamine, 97%