- CH domains revisited.
CH domains revisited.
FEBS letters (1998-08-26)
T Stradal, W Kranewitter, S J Winder, M Gimona
PMID9708889
ABSTRACT
A sequence motif of about 100 amino acids, termed the 'calponin homology domain' has been suggested to confer actin binding to a variety of cytoskeletal and signalling molecules. Here we analyse and compare the sequences of all calponin homology domain-containing proteins identified to date. We propose that single calponin homology domains do not confer actin-binding per se and that the actin-binding motifs of cross-linking proteins, which comprise two disparate calponin homology domains, represent a unique protein module.