Merck
  • Assignment of the gene(s) involved in the expression of the proliferation-related Ki-67 antigen to human chromosome 10.
Assignment of the gene(s) involved in the expression of the proliferation-related Ki-67 antigen to human chromosome 10.
Human genetics (1989-10-01)
D M Schonk, H J Kuijpers, E van Drunen, C H van Dalen, A H Geurts van Kessel, R Verheijen, F C Ramaekers
ABSTRACT
The antigen recognized by the monoclonal antibody Ki-67 is a proliferation-related nucleolus-associated constituent used as a marker for cycling cells in tumor diagnosis. Antibody Ki-67 reacts with human proliferating cells, but not with hamster and mouse cells. Expression of the Ki-67 antigen was studied in a panel of human-rodent somatic cell hybrids. The results indicate that a gene involved in the expression of the antigen is located on chromosome 10.