Merck
  • On the spontaneous carboxylation of 1-butyl-3-methylimidazolium acetate by carbon dioxide.
On the spontaneous carboxylation of 1-butyl-3-methylimidazolium acetate by carbon dioxide.
Chemical communications (Cambridge, England) (2011-12-14)
Marcel Besnard, M Isabel Cabaço, Fabián Vaca Chávez, Noël Pinaud, Pedro J Sebastião, João A P Coutinho, Yann Danten
ABSTRACT
The formation of 1-butyl-3-methylimidazolium-2-carboxylate in the mixture of CO(2) with 1-butyl-3-methylimidazolium acetate under mild conditions (298 K, 0.1 MPa) has been put in evidence in the liquid phase using Raman and infrared spectroscopy complemented by DFT calculations and NMR ((1)H, (13)C, (15)N) spectroscopy.
Sigma-Aldrich
1-Butyl-3-methylimidazolium acetate, ≥96.0% (HPLC)