- Assignment of the protein kinase C delta polypeptide gene (PRKCD) to human chromosome 3 and mouse chromosome 14.
Assignment of the protein kinase C delta polypeptide gene (PRKCD) to human chromosome 3 and mouse chromosome 14.
Genomics (1994-01-01)
K Huppi, D Siwarski, J Goodnight, H Mischak
PMID8188219
ABSTRACT
The protein kinase C (pkc) enzymes are a family of serine-threonine protein kinases, each encoded by a distinct and separate gene. The chromosomal locations of human PRKCA, PRKCB, and PRKCG have previously been established. We now report that PRKCD, a novel member of the pkc gene family, maps to human chromosome 3. The chromosomal location of Pkcd has also been determined in the mouse by analysis of recombination frequency in an interspecific panel of backcross mice. We find that the locus encoding pkcd resides proximal to nucleoside phosphorylase (Np-2) and Tcra on mouse chromosome 14 in a region syntenic with human 3p.