Merck
Highly selective C-H functionalization/halogenation of acetanilide.
Journal of the American Chemical Society (2006-06-08)
Xiaobing Wan, Zhongxun Ma, Bijie Li, Keya Zhang, Shaokui Cao, Shiwei Zhang, Zhangjie Shi
ABSTRACT
Highly regioselective C-H functionalization/halogenation of acetanilides to produce ortho-haloacetanilides was catalyzed by Pd(OAc)2 and Cu(OAc) 2 with CuX2 as the halogen source.
Sigma-Aldrich
Acetanilide, purified by sublimation, ≥99.9%
Sigma-Aldrich
Acetanilide, zone-refined, purified by sublimation, ≥99.95%