Merck
  • Synergistic effect of liver X receptor activation and simvastatin on plaque regression and stabilization: an magnetic resonance imaging study in a model of advanced atherosclerosis.
Synergistic effect of liver X receptor activation and simvastatin on plaque regression and stabilization: an magnetic resonance imaging study in a model of advanced atherosclerosis.
European heart journal (2011-05-25)
Chiara Giannarelli, Giovanni Cimmino, Thomas M Connolly, Borja Ibanez, Josè M Garcia Ruiz, Matilde Alique, M Urooj Zafar, Valentin Fuster, Giora Feuerstein, Juan J Badimon
ABSTRACT
The aim of this study was to investigate the effects of liver X receptors (LXRs)-β preferential activation by LXR-623 (WAY-252623), a novel LXRs agonist, on plaque progression/regression in a rabbit model of advanced atherosclerosis. Advanced atherosclerosis was induced in New Zealand White Rabbits (n= 41). At the end of atherosclerosis induction, animals underwent a baseline magnetic resonance imaging (MRI) and were randomized to receive LXR-623 (1.5, 5, or 15 mg/kg/day), simvastatin (5 mg/kg/day), or placebo. The combination of LXR-1.5/simvastatin was also tested. After a final MRI, animals were euthanized and their aortas processed for further analysis. Simvastatin significantly reduced lesion progression (-25%; P< 0.01) in comparison with the placebo group. A similar effect was observed in the LXR-1.5 and -5 groups. A significant regression (16.5%; P< 0.01) of existing atherosclerosis was observed in the LXR-1.5/simvastatin group. Histological and molecular analysis showed plaque stabilization in the animals treated with the LXR-1.5 and -5, and LXR-1.5/simvastatin. The effects of LXR-623 were observed in the presence of a non-significant effect on total-cholesterol, low-density lipoproteins-cholesterol, and triglyceride levels. The results of the present study show that LXR-623 significantly reduces the progression of atherosclerosis and induces plaque regression in combination with simvastatin. These observations could drive future development of novel anti-atherosclerotic therapeutic approaches.
Sigma-Aldrich
WAY-252623, ≥98% (HPLC)